Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071185.1 | XP_020926844.1 | 4274 | 87 | ||
| XM_021071187.1 | XP_020926846.1 | 2388 | 87 | |||
| XM_021071188.1 | XP_020926847.1 | 2443 | 87 | |||
| XM_021071189.1 | XP_020926848.1 | 2452 | 87 | |||
| XM_021071190.1 | XP_020926849.1 | 1741 | 87 | |||
| XM_021071191.1 | XP_020926850.1 | 2671 | 87 | |||
| XM_021071192.1 | XP_020926851.1 | 2507 | 87 | |||
| XM_021071193.1 | XP_020926852.1 | 5657 | 87 | |||
More Information
Set Membership: |
|

NAALADL2
Genomic Map