Assay Details
Gene Symbol: LOC100156277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021073254.1 | XP_020928913.1 | 1556 | 120 | ||
XM_021073255.1 | XP_020928914.1 | 3631 | 120 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
