Assay Details
| Gene Symbol: LOC102162058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076824.1 | XP_020932483.1 | 42 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102162058| Gene Symbol: LOC102162058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076824.1 | XP_020932483.1 | 42 | 88 | ||