Assay Details
| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077840.1 | XP_020933499.1 | 3485 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APCDD1L| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077840.1 | XP_020933499.1 | 3485 | 63 | ||
Set Membership: |
|