Assay Details
| Gene Symbol: LOC110257484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078221.1 | XP_020933880.1 | 2241 | 76 | ||
| XM_021078222.1 | XP_020933881.1 | 2241 | 76 | |||
More Information
Set Membership: |
|

LOC110257484
Genomic Map