Assay Details
| Gene Symbol: LOC110257484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078221.1 | XP_020933880.1 | 2288 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110257484| Gene Symbol: LOC110257484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078221.1 | XP_020933880.1 | 2288 | 94 | ||