Assay Details
Gene Symbol: C18H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078695.1 | XP_020934354.1 | 788 | 86 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C18H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078695.1 | XP_020934354.1 | 788 | 86 |