Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122846.6 | XP_003122894.3 | 1574 | 73 | ||
| XM_013994380.2 | XP_013849834.2 | 1107 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122846.6 | XP_003122894.3 | 1574 | 73 | ||
| XM_013994380.2 | XP_013849834.2 | 1107 | 73 | |||
Set Membership: |
|