Assay Details
| Gene Symbol: STAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123065.5 | XP_003123113.1 | 1217 | 64 | ||
| XM_005661332.3 | XP_005661389.1 | 1290 | 64 | |||
| XM_005661333.3 | XP_005661390.1 | 1209 | 64 | |||
| XM_021084066.1 | XP_020939725.1 | 1214 | 64 | |||
More Information
Set Membership: |
|

STAP2
Genomic Map