Assay Details
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124710.4 | XP_003124758.1 | 1498 | 57 | ||
| XM_005662239.3 | XP_005662296.1 | 1457 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124710.4 | XP_003124758.1 | 1498 | 57 | ||
| XM_005662239.3 | XP_005662296.1 | 1457 | 57 | |||
Set Membership: |
|