Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125691.5 | XP_003125739.2 | 1002 | 75 | ||
| XM_013996914.2 | XP_013852368.1 | 1002 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIRREL1| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125691.5 | XP_003125739.2 | 1002 | 75 | ||
| XM_013996914.2 | XP_013852368.1 | 1002 | 75 | |||