Assay Details
Gene Symbol: FRMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021080497.1 | XP_020936156.1 | 1114 | 68 | ||
XM_021080498.1 | XP_020936157.1 | 1069 | 68 | |||
XM_021080499.1 | XP_020936158.1 | 1987 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
