Assay Details
| Gene Symbol: LOC110257816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080523.1 | XP_020936182.1 | 1823 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110257816| Gene Symbol: LOC110257816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080523.1 | XP_020936182.1 | 1823 | 58 | ||
Set Membership: |
|