Assay Details
| Gene Symbol: CUNH14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081559.1 | XP_020937218.1 | 567 | 97 | ||
| XM_021081560.1 | XP_020937219.1 | 586 | 97 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH14orf180| Gene Symbol: CUNH14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081559.1 | XP_020937218.1 | 567 | 97 | ||
| XM_021081560.1 | XP_020937219.1 | 586 | 97 | |||