Assay Details
| Gene Symbol: LOC100625662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082322.1 | XP_020937981.1 | 705 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100625662| Gene Symbol: LOC100625662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082322.1 | XP_020937981.1 | 705 | 77 | ||