Assay Details
| Gene Symbol: LOC100523114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083173.1 | XP_020938832.1 | 725 | 149 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100523114| Gene Symbol: LOC100523114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083173.1 | XP_020938832.1 | 725 | 149 | ||
Set Membership: |
|