Assay Details
| Gene Symbol: LOC110260022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087566.1 | XP_020943225.1 | 644 | 100 | ||
| XM_021087567.1 | XP_020943226.1 | 749 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110260022| Gene Symbol: LOC110260022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087566.1 | XP_020943225.1 | 644 | 100 | ||
| XM_021087567.1 | XP_020943226.1 | 749 | 100 | |||