Assay Details
| Gene Symbol: C3H16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088380.1 | XP_020944039.1 | 75 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3H16orf90| Gene Symbol: C3H16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088380.1 | XP_020944039.1 | 75 | 86 | ||