Assay Details
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089188.1 | XP_020944847.1 | 1398 | 67 | ||
| XM_021089189.1 | XP_020944848.1 | 1398 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: JPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089188.1 | XP_020944847.1 | 1398 | 67 | ||
| XM_021089189.1 | XP_020944848.1 | 1398 | 67 | |||
Set Membership: |
|