Assay Details
| Gene Symbol: LOC100513083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090821.1 | XP_020946480.1 | 8328 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100513083| Gene Symbol: LOC100513083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090821.1 | XP_020946480.1 | 8328 | 74 | ||