Assay Details
| Gene Symbol: LOC110260590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091246.1 | XP_020946905.1 | 209 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110260590| Gene Symbol: LOC110260590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091246.1 | XP_020946905.1 | 209 | 82 | ||