Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091590.1 | XP_020947249.1 | 1657 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC3L| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091590.1 | XP_020947249.1 | 1657 | 105 | ||
Set Membership: |
|