Assay Details
| Gene Symbol: LOC110260994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094471.1 | XP_020950130.1 | 966 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110260994| Gene Symbol: LOC110260994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094471.1 | XP_020950130.1 | 966 | 71 | ||
Set Membership: |
|