Assay Details
| Gene Symbol: LOC102164418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096717.1 | XP_020952376.1 | 1099 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102164418| Gene Symbol: LOC102164418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096717.1 | XP_020952376.1 | 1099 | 92 | ||
Set Membership: |
|