Assay Details
| Gene Symbol: WTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097104.1 | XP_020952763.1 | 754 | 73 | ||
| XM_021097105.1 | XP_020952764.1 | 754 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: WTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097104.1 | XP_020952763.1 | 754 | 73 | ||
| XM_021097105.1 | XP_020952764.1 | 754 | 73 | |||
Set Membership: |
|