Assay Details
Gene Symbol: C6H19orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097417.1 | XP_020953076.1 | 2922 | 66 | ||
XM_021097418.1 | XP_020953077.1 | 1153 | 66 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C6H19orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097417.1 | XP_020953076.1 | 2922 | 66 | ||
XM_021097418.1 | XP_020953077.1 | 1153 | 66 |