Assay Details
Gene Symbol: C6H19orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097417.1 | XP_020953076.1 | 3024 | 63 | ||
XM_021097418.1 | XP_020953077.1 | 1255 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
