Assay Details
| Gene Symbol: MEIOSIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097552.1 | XP_020953211.1 | 1634 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIOSIN| Gene Symbol: MEIOSIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097552.1 | XP_020953211.1 | 1634 | 71 | ||