Assay Details
| Gene Symbol: GREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098305.1 | XP_020953964.1 | 279 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021098305.1 transcripts.
Set Membership: |
|

GREM1
Genomic Map