Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099551.1 | XP_020955210.1 | 244 | 102 | ||
| XM_021099560.1 | XP_020955219.1 | 244 | 102 | |||
More Information
Set Membership: |
|

SERPINB13
Genomic Map