Assay Details
| Gene Symbol: LOC100512793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099690.1 | XP_020955349.1 | 3686 | 49 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100512793| Gene Symbol: LOC100512793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099690.1 | XP_020955349.1 | 3686 | 49 | ||
Set Membership: |
|