Assay Details
| Gene Symbol: LOC100155488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100051.1 | XP_020955710.1 | 216 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100155488| Gene Symbol: LOC100155488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100051.1 | XP_020955710.1 | 216 | 97 | ||
Set Membership: |
|