Assay Details
| Gene Symbol: GCM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100173.1 | XP_020955832.1 | 446 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GCM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100173.1 | XP_020955832.1 | 446 | 78 | ||