Assay Details
Gene Symbol: TAS2R9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126499.2 | XP_003126547.1 | 822 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
