Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130104.4 | XP_003130152.1 | 669 | 77 | ||
| XM_013989532.2 | XP_013844986.1 | 669 | 77 | |||
| XM_021063189.1 | XP_020918848.1 | 669 | 77 | |||
| XM_021063190.1 | XP_020918849.1 | 669 | 77 | |||
| XM_021063191.1 | XP_020918850.1 | 669 | 77 | |||
More Information
Set Membership: |
|

FMO2
Genomic Map