Assay Details
| Gene Symbol: ACVR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133418.5 | XP_003133466.3 | 1028 | 67 | ||
| XM_013984144.2 | XP_013839598.1 | 950 | 67 | |||
| XM_013984145.2 | XP_013839599.1 | 873 | 67 | |||
More Information
Set Membership: |
|

ACVR1C
Genomic Map