Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134060.5 | XP_003134108.1 | 794 | 68 | ||
| XM_021076941.1 | XP_020932600.1 | 797 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134060.5 | XP_003134108.1 | 794 | 68 | ||
| XM_021076941.1 | XP_020932600.1 | 797 | 68 | |||
Set Membership: |
|