Assay Details
Gene Symbol: CXHXorf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135428.3 | XP_003135476.1 | 2072 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135428.3 transcripts.
Set Membership: |
![]() ![]() |
