Assay Details
| Gene Symbol: LOC100623541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354701.4 | XP_003354749.1 | 184 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003354701.4 transcripts.
Set Membership: |
|

LOC100623541
Genomic Map