Assay Details
| Gene Symbol: LOC100623541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354701.4 | XP_003354749.1 | 364 | 71 | ||
| XM_013995890.2 | XP_013851344.1 | 283 | 71 | |||
| XM_013995891.2 | XP_013851345.1 | 436 | 71 | |||
| XM_013995892.2 | XP_013851346.1 | 320 | 71 | |||
| XM_013995893.2 | XP_013851347.1 | 254 | 71 | |||
| XM_013995894.2 | XP_013851348.1 | 392 | 71 | |||
More Information
Set Membership: |
|

LOC100623541
Genomic Map