Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355900.4 | XP_003355948.1 | 1335 | 58 | ||
| XM_021094359.1 | XP_020950018.1 | 1338 | 58 | |||
| XM_021094360.1 | XP_020950019.1 | 959 | 58 | |||
More Information
Set Membership: |
|

ACP7
Genomic Map