Assay Details
| Gene Symbol: NSUN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356901.4 | XP_003356949.1 | 1838 | 77 | ||
| XM_005666617.2 | XP_005666674.1 | 1789 | 77 | |||
| XM_021101136.1 | XP_020956795.1 | 1783 | 77 | |||
More Information
Set Membership: |
|

NSUN7
Genomic Map