Assay Details
| Gene Symbol: LOC100624788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356958.4 | XP_003357006.2 | 863 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100624788| Gene Symbol: LOC100624788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356958.4 | XP_003357006.2 | 863 | 109 | ||
Set Membership: |
|