Assay Details
| Gene Symbol: LOC100624788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356958.4 | XP_003357006.2 | 1144 | 105 | ||
| XM_005656509.3 | XP_005656566.2 | 1066 | 105 | |||
More Information
Set Membership: |
|

LOC100624788
Genomic Map