Assay Details
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003130445.3 | XP_003130493.1 | 1221 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
