Assay Details
| Gene Symbol: LRRC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132076.6 | XP_003132124.1 | 1517 | 82 | ||
| XM_005669310.3 | XP_005669367.1 | 1364 | 82 | |||
| XM_005669311.3 | XP_005669368.1 | 1373 | 82 | |||
| XM_005669312.3 | XP_005669369.1 | 840 | 82 | |||
| XM_013981425.2 | XP_013836879.1 | 1596 | 82 | |||
| XM_021071452.1 | XP_020927111.1 | 2930 | 82 | |||
| XM_021071453.1 | XP_020927112.1 | 1449 | 82 | |||
More Information
Set Membership: |
|

LRRC3B
Genomic Map