Assay Details
| Gene Symbol: C13H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981667.2 | XP_013837121.1 | 5427 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C13H3orf62| Gene Symbol: C13H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981667.2 | XP_013837121.1 | 5427 | 76 | ||
Set Membership: |
|