Assay Details
| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988516.2 | XP_013843970.1 | 420 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPEP2NB| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988516.2 | XP_013843970.1 | 420 | 69 | ||
Set Membership: |
|