Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360486.4 | XP_003360534.2 | 615 | 82 | ||
| XM_003360487.4 | XP_003360535.2 | 615 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360486.4 | XP_003360534.2 | 615 | 82 | ||
| XM_003360487.4 | XP_003360535.2 | 615 | 82 | |||
Set Membership: |
|