Assay Details
| Gene Symbol: LRRC56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082550.1 | XP_020938209.1 | 669 | 82 | ||
| XM_021082551.1 | XP_020938210.1 | 719 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LRRC56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082550.1 | XP_020938209.1 | 669 | 82 | ||
| XM_021082551.1 | XP_020938210.1 | 719 | 82 | |||
Set Membership: |
|